Få mere at vide
Brodimoprim-d6, TRC
Artikelnummer. 30339996
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30339996 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339996 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6781021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Brodimoprim-d6
CAS 1346599-93-6
Molekylær formel C13H9D6BrN4O2
Mængde 1 mg
Synonym 5-[(4-Bromo-3,5-(dimethoxy-d6)phenyl)methyl]-2,4-pyrimidinediamine, 5-(4-Bromo-3,5-(dimethoxy-d6)benzyl)-2,4-pyrimidinediamine, Hyprim-d6, Ro 10-5970-d6, Ro 105970-d6, Unitrim-d6
InChI formel InChI=1S/C13H15BrN4O2/c1-19-9-4-7(5-10(20-2)11(9)14)3-8-6-17-13(16)18-12(8)15/h4-6H,3H2,1-2H3,(H4,15,16,17,18)/i1D3,2D3
SMIL [2H]C([2H])([2H])Oc1cc(CC2=CNC(=N)NC2=N)cc(OC([2H])([2H])[2H])c1Br
IUPAC navn 5-[[4-bromo-3,5-bis(trideuteriomethoxy)phenyl]methyl]-1H-pyrimidine-2,4-diimine
Molekylvægt (g/mol) 345.22
Formel vægt 344.0755