Få mere at vide
Brimonidine-d4 D-Tartrate, TRC
Artikelnummer. 30340418
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30340418 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340418 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6775321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Brimonidine-d4 D-Tartrate
CAS 1316758-27-6
Molekylær formel C15H12D4BrN5O6
Mængde 1 mg
Synonym 5-Bromo-N-(4,5-dihydro-1H-imidazol-d4-2-yl)-6-quinoxalinamine D-Tartrate, 5-Bromo-6-(2-imidazolin-d4-2-ylamino)quinoxaline D-Tartrate, UK-14304-18-d4, AGN-190342-LF-d4, Alphagan-d4
InChI formel InChI=1S/C11H10BrN5.C4H6O6/c12-9-7(17-11-15-5-6-16-11)1-2-8-10(9)14-4-3-13-8;5-1(3(7)8)2(6)4(9)10/h1-4H,5-6H2,(H2,15,16,17);1-2,5-6H,(H,7,8)(H,9,10)/t;1-,2-/m.0/s1/i5D2,6D2;
SMIL BrC1=C2C(N=CC=N2)=CC=C1NC3=NC([2H])([2H])C([2H])([2H])N3.O[C@@H]([C@H](O)C(O)=O)C(O)=O
IUPAC navn 5-bromo-N-(4,4,5,5-tetradeuterio-1H-imidazol-2-yl)quinoxalin-6-amine;(2S,3S)-2,3-dihydroxybutanedioic acid
Molekylvægt (g/mol) 446.25