Få mere at vide
Brilliant Sulfaflavine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acid Yellow 7 |
| CAS | 2391-30-2 |
| Molekylær formel | C19 H13 N2 O5 S . Na |
| Mængde | 100 mg |
| Synonym | Lissamine Flavine FFS, Acid New Yellow GMF, Acid Yellow 7, Brilliant Acid Flavine 10J, Brilliant Acid Yellow 8G, Brilliant Sulfo Flavin FFA, Brilliant Sulfo Flavine, Brilliant Sulfo Flavine FF, Brilliant Sulfo Flavine FFA, Brilliant Sulfoflavin, Brilliant Sulfoflavin FF, Brilliant Sulphaflavine, Brilliant Sulpho Flavine FF, Brilliant Sulpho Flavine FFA, Brilliant Sulfaflavine, C.I. 56205, C.I. Acid Yellow 7, Dyacid Yellow FF, Fenazo Yellow XX, Kayaku Acid Brilliant Flavine FF, Lissamine Flavine, Lissamine Flavine FF, Lissamine Yellow FF, Naphthazine Flavine FF, Sulfacid Brilliant Yellow 10J, Sulfaflavine, Triacid Flavine FF, 6-Amino-2,3-dihydro-1,3-dioxo-2-p-tolyl-1H-benz[de]isoquinoline-5-sulfonic Acid Sodium Salt, 6-Amino-2,3-dihydro-2-(4-methylphenyl)-1,3-dioxo-1H-benz[de]isoquinoline-5-sulfonic Acid Monosodium Salt |
| InChI formel | InChI=1S/C19H14N2O5S.Na/c1-10-5-7-11(8-6-10)21-18(22)13-4-2-3-12-16(13)14(19(21)23)9-15(17(12)20)27(24,25)26;/h2-9H,20H2,1H3,(H,24,25,26);/q;+1/p-1 |
| SMIL | [Na+].Cc1ccc(cc1)N2C(=O)c3cccc4c(N)c(cc(C2=O)c34)S(=O)(=O)[O-] |
| IUPAC navn | sodium;6-amino-2-(4-methylphenyl)-1,3-dioxobenzo[de]isoquinoline-5-sulfonate |
| Molekylvægt (g/mol) | 404.37 |
| Formel vægt | 404.0443 |