Få mere at vide
Brexpiprazole-d8, TRC
Artikelnummer. 30339482
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30339482 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30339482 Leverandør Toronto Research Chemicals Leverandørnr. TRCB67738725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Brexpiprazole-d8
CAS 1427049-19-1
Renhedsgrad noter HPLC
Molekylær formel C25H19D8N3O2S
Mængde 25 mg
Synonym 7-[4-(4-(Benzo[b]thien-4-yl)-piperazin-1-yl)butoxy]-1H-quinolin-2-one-d8, OPC 34712-d8, 2(1H)-Quinolinone, 7-[4-(4-benzo[b]thien-4-yl-1-piperazinyl)butoxy-1,1,2,2,3,3,4,4-d8]-, 7-[4-(4-Benzo[b]thien-4-yl-1-piperazinyl)butoxy-1,1,2,2,3,3,4,4-d8]-2(1H)-quinolinone, 7-[4-(4-(Benzo[b]thien-4-yl)-piperazin-1-yl)-1,1,2,2,3,3,4,4-octadeuteriobutoxy]-1H-quinolin-2-one, Brexpiprazole-D8 (butoxy-D8)
InChI formel InChI=1S/C25H27N3O2S/c29-25-9-7-19-6-8-20(18-22(19)26-25)30-16-2-1-11-27-12-14-28(15-13-27)23-4-3-5-24-21(23)10-17-31-24/h3-10,17-18H,1-2,11-16H2,(H,26,29)/i1D2,2D2,11D2,16D2
SMIL [2H]C([2H])(Oc1ccc2C=CC(=O)Nc2c1)C([2H])([2H])C([2H])([2H])C([2H])([2H])N3CCN(CC3)c4cccc5sccc45
IUPAC navn 7-[4-[4-(1-benzothiophen-4-yl)piperazin-1-yl]-1,1,2,2,3,3,4,4-octadeuteriobutoxy]-1H-quinolin-2-one
Molekylvægt (g/mol) 441.62
Formel vægt 441.2326
Procent renhed >95