missing translation for 'onlineSavingsMsg'
Få mere at vide

Brefeldin A, MedChemExpress

Artikelnummer. 16469637
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16469637 5 mg
16427136 100 mg
16417098 10 mg
16433137 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16469637 Leverandør Medchem Express Leverandørnr. HY16592

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Brefeldin A (BFA) is a lactone antibiotic and a specific inhibitor of protein trafficking. Brefeldin A blocks the transport of secreted and membrane proteins from endoplasmic reticulum to Golgi apparatus. Brefeldin A is also an autophagy and mitophagy inhibitor. Brefeldin A is a CRISPR/Cas9 activator. Brefeldin A inhibits HSV-1 and has anti-cancer activity.

Brefeldin A (BFA) is a lactone antibiotic and a specific inhibitor of protein trafficking. Brefeldin A blocks the transport of secreted and membrane proteins from endoplasmic reticulum to Golgi apparatus. Brefeldin A is also an autophagy and mitophagy inhibitor. Brefeldin A is a CRISPR/Cas9 activator. Brefeldin A inhibits HSV-1 and has anti-cancer activity.

Tekniske data

Kemisk navn eller materiale Brefeldin A
Fysisk form Solid
CAS 20350-15-6
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C16H24O4
Til brug med (applikation) Cancer-programmed cell death
Synonym BFA Cyanein Decumbin
Opløselighedsinformation DMSO : 100 mg/mL (356.68 mM; Need ultrasonic) ∣Ethanol : 11.11 mg/mL (39.63 mM; Need ultrasonic)
SMIL O[C@@H]1C[C@]2([H])[C@@](/C=C/CCC[C@H](C)OC(/C=C/[C@H]2O)=O)([H])C1
Molekylvægt (g/mol) 280.36
Formel vægt 280.36
Procent renhed 95.0%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.