missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRD7-IN-1, MedChemExpress
Description
BRD7-IN-1, a modified derivative of BI7273 (BRD7/9 inhibitor), binds to a VHL ligand via a linker to form a PROTAC VZ185 (VZ185 against BRD7/9 with DC50s of 4.5 and 1.8 nM, respectively).
Specifications
Specifications
| Kemisk navn eller materiale | BRD7-IN-1 |
| CAS | 2448414-48-8 |
| Farve | Yellow |
| Renhedsgrad noter | Research |
| Molekylær formel | C22H28Cl2N4O3 |
| Mængde | 5 mg |
| Opløselighedsinformation | H2O : 100 mg/mL (213.95 mM; Need ultrasonic) ∣DMSO : 30 mg/mL (64.19 mM; Need ultrasonic) |
| SMIL | CN1C=C(C2=CC(OC)=C(CN3CCNCC3)C(OC)=C2)C(C=CN=C4)=C4C1=O.Cl.Cl |
| Molekylvægt (g/mol) | 467.39 |
| Formel vægt | 467.39 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?