Få mere at vide
BRD 6989
Mængde:
10 mg
50 mg
Beskrivelse
Binds and inhibits Cdk8 in complex with cyclin C (IC50 ~ 200 nM). Enhances IL-10 production in activated human and murine macrophages and dendritic cells (EC50 is approximately 1 μM for IL-10 production in bone marrow-derived dendritic cells). Also inhibits CDK19 in complex with cyclin C.
Tekniske data
Tekniske data
| CAS | 642008-81-9 |
| Synonym | 2-Amino-5,6,7,8-tetrahydro-6-methyl-4-(3-pyridinyl)-3-quinolinecarbonitrile |
| Mængde | 50 mg |
| Molekylær formel | C16H16N4 |
| Formel vægt | 264.33 |
| Renhed | 98% |