Få mere at vide
Cole-Parmer™ Lige kompressionsunion af messing
Slangestørrelse:
1/16 tommer.
1/2 tomme
1/4 tommer
1/8 tomme
Pakningsstørrelse:
Hver
Beskrivelse
- Very good chemical resistance.
- Compression fittings work well with rigid tubing such as PTFE or polypropylene.
- A compression fitting has a nut that compresses rigid tubing against fitting body for high-pressure applications.
- Compression connector family is ideal when working with rigid tubing.
- When selecting your fittings, consider your tubing type, material compatibility and temperature and pressure conditions.
Tekniske data
Tekniske data
| Autoklaverbar | Not Autoclavable |
| Materiale | Brass |
| Min. Driftstemperatur (engelsk) | -85°F |
| Min. Driftstemperatur (metrisk) | -65°C |
| Slangestørrelse | 1/2 tomme |
| Maksimalt tryk | 400 psi |
| Maks. Driftstemperatur (engelsk) | 482°F |
| Maks. Driftstemperatur (metrisk) | 250°C |
| Produkttype | Straight Compression Union |