Learn More
Branson Ultrasonics™ Ultrasonic Cleaning Bath Accessory, Covers
For use with Bransonic cleaning baths
Brand: Branson Ultrasonics™ 100-032-510
Additional Details : Weight : 0.40000kg
Beschreibung
For use with Bransonic cleaning baths
- Stainless steel lid
Spezifikation
Lid | |
Ultrasonic Bath, Model 5800 | |
Helps position beakers securely in bath | |
Plastic |