Learn More
BRAND™ 384-Well PCR Plates
Made with polypropylene material, these plates are resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO and is compatible with most robotic samplers and automated liquid handling systems.
2945.00 DKK
Specifications
Format | Standard |
---|---|
Array | 16 x 24 |
Autoclavable | Autoclavable |
Skirt Style | Full |
Material | Polypropylene |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
10446593
|
BRAND™
781347 |
Full skirt |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Non-sterile, 0.3mL BRAND™ 384-well deep well plate has a square bottom and is manufactured from high-quality virgin polypropylene
- Excellent for high throughput screening (HTS), assays requiring a mother plate, cell and tissue culture, sample storage, and more
- Plate is resistant to a wide variety of chemicals, including phenols, chloroform, and DMSO and is compatible with most robotic samplers and automated liquid handling systems
- Plate withstands temperatures as low as -121ºC, making it an excellent choice for cold-room applications and cryo-storage
- PP plate is autoclavable at 250ºF (121ºC) for 20 minutes
- Wells are labeled in a standard alpha-numeric pattern to simplify sample identification, and have notched corners to facilitate orientation
Specifications
Standard | |
Autoclavable | |
Polypropylene | |
qPCR |
16 x 24 | |
Full | |
384 | |
50/Pk. |