Learn More
Bradykinin (1-5), Thermo Scientific Chemicals
For cellular and molecular biology applications
Brand: Thermo Scientific Alfa Aesar J63233.MB
Additional Details : Weight : 0.00025kg
Description
Biologically active fragment of bradykinin
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
HPLC | |
Powder | |
572.67 | |
25 mg | |
H-Arg-Pro-Pro-Gly-Phe-OH |
-30°C to -10°C | |
Biologically active fragment of bradykinin | |
Bradykinin (1-5) | |
RUO - research use only |
RUO – Research Use Only