Learn More
Trajan™ BP5 GC Capillary Column
A general purpose phase used for a wide range of applications
Brand: Trajan™ 054167
This item is not returnable.
View return policy
Description
- Excellent general purpose column
- Low bleed
- Non-polar
- High temperature
- Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
320/340°C | |
5% Phenyl/95% Dimethyl Polysiloxane | |
Drugs of Abuse Analysis, Pesticide Analysis, Herbicides Analysis, Drugs of Abuse, Hydrocarbons | |
0.25 ÎĽm | |
Forte™ BP5 | |
Fused Silica |
0.22 mm | |
G36, G41 | |
-60°C to +320/340°C | |
12 m | |
Non polar |
Product Content Correction
Product Title
Trajan™ BP5 GC Capillary Column > Film thickness: 0.25um; I.D.: 0.22mm; Length: 12m
Spot an opportunity for improvement?Share a Content Correction