Learn More
Trajan™ BP1 GC Capillary Columns
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
Brand:  Trajan™ 054046
 View more versions of this product
Product Code. 10298760
  • 3225.00 DKK / Each
Only null left
Estimated Shipment: 14-07-2025
Description
Suitable for analysis of hydrocarbons, aromatics, pesticides, phenol, herbicides, and amines. Trajan Scientific and Medical™ BP1 CG Capillary Columns are constructed of 100% Dimethyl Polysiloxane.
  • Classic cross-linked dimethyl polysiloxane technology
  • Low bleed
  • Non-polar
  • Ideal for analyses such as hydrocarbons, aromatics, pesticides and amines
Specifications
Specifications
360°C
0.22 mm
-60°C to +360°C
12 mm
BP1 GC Capillary Column
100% Dimethyl Polysiloxane
0.25 mm
1 Each
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Trajan™ BP1 GC Capillary Columns > Diameter Outer: ; Length:12mm