Learn More
Eppendorf™ Box of Eppendorf Quality™ epT.I.P.S.™ Pipette Tips
Eppendorf™ Box of Eppendorf Quality™ epT.I.P.S.™ Pipette Tips are the perfect combination that results in reduced-force tip attachment and ejection with complete sealing.
Brand:  Eppendorf™ 0030073657
Additional Details : Weight : 0.54000kg
 View more versions of this product
Product Code. 15851505
  • 270.00 DKK / Pack of 96
Estimated Shipment
12-04-2019
Add to basket
Description
Description
  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Boxes and trays are autoclavable at 121°C for subsequent use
Specifications
Specifications
Box
50 to 1250μL
Natural
Green
epT.I.P.S.™ Box
Eppendorf Quality™
1 reusable box x 96 tips
Non-Filtered
Green
Yes
EN ISO 8655
Videos