missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Box of Eppendorf Quality™ epT.I.P.S.™ Pipette Tips

Eppendorf™ Box of Eppendorf Quality™ epT.I.P.S.™ Pipette Tips are the perfect combination that results in reduced-force tip attachment and ejection with complete sealing.

Brand:  Eppendorf™ 0030073150

Additional Details : Weight : 0.48700kg

 View more versions of this product

Product Code. 15851485

  • 310.00 DKK / Pack of 96


Estimated Shipment
12-06-2019
Add to basket

Description

Description

  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Boxes and trays are autoclavable at 121°C for subsequent use
Specifications

Specifications

Box
0.25 to 2.5mL
Natural
Red
epT.I.P.S.™ Box
EN ISO 8655
1 reusable box x 48 tips
Non-Filtered
Red
Yes
115mm
Eppendorf Quality™
Videos