Få mere at vide
Bovine Serum Albumin Antibody (ALB/398), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76991B
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Bovine Serum Albumin Monoclonal antibody specifically detects Bovine Serum Albumin in Bovine, Canine (Negative), Guinea Pig (Negative), Human (Negative), Rabbit (Negative) samples. It is validated for ELISA
Tekniske data
| Bovine Serum Albumin | |
| Monoclonal | |
| Biotin | |
| albumin, BSA, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
| Purified Bovine Serum Albumin (Uniprot: P02769) | |
| 0.1 mL | |
| Apoptosis, Cancer, Golgi Apparatus Markers, Stem Cell Markers | |
| 280717 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| ALB/398 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Bovine, Canine (Negative), Guinea Pig (Negative), Human (Negative), Rabbit (Negative) | |
| Purified |