Learn More
Eppendorf™ Bottle Thread Adapter
Description
- ETFE
- From GL 32 to GL/S 28
Specifications
Specifications
| Product Type | Bottle Thread Adapter |
| Material | ETFE |
| Thread Size | GL 32 to GL/S 28 |