Få mere at vide
Bosentan-d4, TRC
Artikelnummer. 30329426
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30329426 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30329426 Leverandør Toronto Research Chemicals Leverandørnr. TRCB67590225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bosentan-d4
CAS 1065472-77-6
Renhedsgrad noter HPLC
Molekylær formel C27H25D4N5O6S
Mængde 25 mg
Synonym 4-(1,1-Dimethylethyl)-N-[6-(2-hydroxyethoxy-d4)-5-(2-methoxyphenoxy)[2,2’-bipyrimidin]-4-yl]benzenesulfonamide, p-tert-Butyl-N-[6-(2-hydroxyethoxy-d4)-5- (o-methoxyphenoxy)-2-(2-pyrimidinyl)-4-pyrimidinyl]benzenesulfonamide, Actelion-d4, Ro 47-0203-d4, Ro 47-0203/039-d4, Tracleer-d4
InChI formel InChI=1S/C27H29N5O6S/c1-27(2,3)18-10-12-19(13-11-18)39(34,35)32-23-22(38-21-9-6-5-8-20(21)36-4)26(37-17-16-33)31-25(30-23)24-28-14-7-15-29-24/h5-15,33H,16-17H2,1-4H3,(H,30,31,32)/i16D2,17D2
SMIL OC([2H])([2H])C([2H])([2H])OC1=C(OC2=CC=CC=C2OC)C(NS(C3=CC=C(C(C)(C)C)C=C3)(=O)=O)=NC(C4=NC=CC=N4)=N1
IUPAC navn 4-tert-butyl-N-[5-(2-methoxyphenoxy)-2-pyrimidin-2-yl-6-(1,1,2,2-tetradeuterio-2-hydroxyethoxy)pyrimidin-4-yl]benzenesulfonamide
Molekylvægt (g/mol) 555.64
Procent renhed >95