Få mere at vide
Bortezomib, 98%
Artikelnummer. 15453159
Skift visning
Mængde:
5 mg
10 mg
25 mg
Pakningsstørrelse:
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
15453159 5 mg 5mg
15463159 10 mg 10mg
15473159 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 15453159 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. J60378.#0
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Bortezomib (CAS # 179324-69-7) is an inhibitor of protein degradation pathways. | CAS: 179324-69-7 | C19H25BN4O4 | 384.243 g/mol
A potent, highly selective and reversible inhibitor of the 20S proteasome, Bortezomib is used as an antineoplastic agent, which controls the growth of cancer cells. It is also useful in the treatment of multiple myeloma.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Bortezomib is an antineoplastic compound derived from boronic acid
• Bortezomib reversibly inhibits the 26S proteostome that maintains cell cycle pathways related to growth and division
• Bortezomib is an inhibitor of NF-κB, a factor that is constituitively active in some cancers
Application
• Bortezomib is suitable for use in the research of multiple myeloma and lymphomas, and for use in the research of protein degradation pathways
• Bortezomib is intended for in vitro and biochemical applications
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 179324-69-7
Molekylær formel C19H25BN4O4
Molekylvægt (g/mol) 384.243
MDL nummer MFCD09056737
InChI nøgle GXJABQQUPOEUTA-RDJZCZTQSA-N
Synonym bortezomib, velcade, bortezomib ps-341, dpba, pyz-phe-boroleu, unii-69g8bd63pp, velcade tn, bortezomib velcade, 1r-3-methyl-1-2s-1-oxo-3-phenyl-2-pyrazinylcarbonyl amino propyl amino butyl boronic acid, 1r-3-methyl-1-2s-3-phenyl-2-pyrazine-2-carbonylamino propanoyl amino butyl boronic acid
PubChem CID 387447
ChEBI CHEBI:52717
IUPAC navn [(1R)-3-methyl-1-[[(2S)-3-phenyl-2-(pyrazine-2-carbonylamino)propanoyl]amino]butyl]boronic acid
SMIL B(C(CC(C)C)NC(=O)C(CC1=CC=CC=C1)NC(=O)C2=NC=CN=C2)(O)O
Tekniske data
Kemisk navn eller materiale Bortezomib
Smeltepunkt 139°C to 143°C
Farve Brown to White
Mængde 5 mg
FN nummer UN3249
Merck Index 14,1351
Opløselighedsinformation Soluble in DMSO or ethanol
Formel vægt 384.24
Procent renhed 99%
Fysisk form Powder