Learn More
Quickfit™ Borosilicate Glass Stopcock Adapter with Sockets
Control flow rate. QuickFit™ borosilicate glass stopcock adater with cone comes with a GP Rotoflo™ stopcock.
Brand: Quickfit MF14/2/SC
712.50 DKK valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Additional Details : Weight : 0.03000kg
Description
- With GP Rotaflo™ PTFE stopcock providing fine flow control and positive closure
- Detachable plastic screwthread connectors allow rubber hose etc. , to be fitted easily and safely
Specifications
Stopcock Adapter | |
Borosilicate Glass | |
19/26 | |
Sockets |