Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
Brand:  Lenz Laborglasinstrumente™ 04016149
 View more versions of this product
Product Code. 10110551
  • 834.00 DKK / Pack of 2
Only null left
Estimated Shipment: 11-07-2025
Description
  • With NS socket and PE stopper
  • with PTFE plug
  • with screwthread retaining device
  • with scale (approx. volume /ml)
  • material: borosilicate glass 3.3
Specifications
Specifications
250 mL
Borosilicate Glass
2Pack
Polyethylene
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale > Capacity: 250mL