Learn More
Wheaton™ Borosilicate Glass Mortar
1190.00 DKK - 1270.00 DKK
Specifications
Autoclavable | Yes |
---|---|
Material | Borosilicate Glass |
Product Type | Mortar |
For Use With (Equipment) | For Tissue Grinder |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12411471
|
Wheaton™
358010 |
15mL |
1270.00 DKK
Pack of 2 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10446341
|
Wheaton™
358008 |
10mL |
1270.00 DKK
Pack of 2 |
Estimated Shipment: 09-10-2024
Log in to see stock. |
|||||
10534323
|
Wheaton™
358004 |
2mL |
1205.00 DKK
Pack of 2 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
10749435
|
Wheaton™
358006 |
5mL |
1190.00 DKK
Pack of 2 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
|||||
Specifications
Yes | |
Borosilicate Glass | |
Mortar | |
For Tissue Grinder |