Learn More
Wheaton™ Borosilicate Glass Mortar
Brand: Wheaton™ 358008
1206.50 DKK valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.15060kg
Specifications
Yes | |
Borosilicate Glass | |
For Tissue Grinder |
10mL | |
Mortar |