Learn More
Quickfit™ Borosilicate Glass Markham Distillation Assembly
Product Code. 12498786
Change view
Click to view available options
Material:
Borosilicate Glass
Unit Size:
Each
Product Code. Material Unit Size
12498786 Borosilicate Glass Each
1 options
Product Code. 12498786 Supplier Quickfit™ Supplier No. 46MC
  • ONLINE EXCLUSIVE PRICE
    4115.00 DKK
    3194.51 DKK / Pakke med 1
    Spare 920.49 DKK
    22% Slukket
Forventet forsendelse: 12-04-2027
Quickfit™ Borosilicate Glass Markham Distillation Assembly is used for volatile fatty acid tests and can be used to carry out a large number of distillations in rapid successioin
  • Complies with BS1428: Part B2
  • Enables a large number of distillations to be carried out in rapid succession
  • Comprises Markham still body, Liebig condenser and connecting joint clip
  • Still has evaporator vessel enclosed in a steam jacket
  • When connected to a suitable steam supply the steam passes through the sample to promote distillation
  • When steam supply is shut off the apparatus automatically sucks back any residue in the vessel thus preparing itself for the next determination
Specifications
For Use With (Application) Enables a large number of distillations to be carried out in rapid succession
Type Markham distillation assembly
Material Borosilicate Glass
Certifications/Compliance BS 1428: Part B2