Learn More
Kimble Chase™ Borosilicate Glass Vials with Caps
Kimble Chase™ Borosilicate Glass Vials with Caps are suitable for a variety of storage and transportation needs.
Brand:  Kimble Chase™ 60941A-40
Additional Details : Weight : 2.00000kg
 View more versions of this product
Product Code. 13131106
  • 1241.00 DKK / Pack of 72
Estimated Shipment
24-09-2021
Add to basket
Description
Description
  • Clear, screw thread sample vials
  • Available in different sizes and material strengths
  • Black phenolic closure available with different liner types
Specifications
Specifications
28 mm
40 mL
72
98 mm
Borosilicate Glass (Type 1 Class A)
GPI 24-400
Black Phenolic Cap with Cone-shaped Polyethylene Liner
Videos
SDS
missing translation for 'documents'