Få mere at vide
BRAND™ BLAUBRAND™ Compact Automatic Burette with Glass Bottle
Shop alle Brand produkter
Kapacitet (metrisk):
10 ml
50 ml
Pakningsstørrelse:
Hver
Beskrivelse
- Borosilicate glass construction
- Class AS, DE-M marking
- Graduations in blue
- Detachable PTFE stopcock.
- DIN EN ISO 385
- Calibrated to deliver (TD, Ex).
- All BLAUBRAND™ automatic burettes are supplied with a batch number and an accompanying batch certificate in the original packaging. On request, they are available also with an individual certificate, USP individual certificate or DAkkS calibration certificate.
- Includes Burette tube with Schellbach stripe and automatic zeroing, stopcock with precision tip, filling tube (PVC, transparent), pumphead and 2000mL soda-lime glass bottle
- Supports, burette clamps and rubber bellows sold separately
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 50 ml |
| Længde (metrisk) | 790 mm |
| Materiale | Borosilicate Glass |
| Gradueringer | 0.1 mL |
| Inkluderer | Glass Bottle |
| Stophane materiale | PTFE |
| Tolerance | ±0.05 mL |
| Type | Automatic Buret |
| Klasse | Class AS |
| Certificeringer/compliance | DIN EN ISO 385, USP |