Learn More
Boron Standard Metal Solution 1000ppm, for ICP, Solution Aqueous, Fisher Chemical
Click to view available options
Quantity:
500 mL
Packaging:
HDPE plastic bottle
Specifications
Packaging | HDPE plastic bottle |
Quantity | 500 mL |
Color | Colorless |
MDL Number | 134034 |
Physical Form | Liquid |