Learn More
Boron for AAS 1g/L H2O matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Brand: Fisher Chemical B005100
Additional Details : Weight : 0.10000kg
Packaging | Bottle |
---|---|
Quantity | 100mL |
Chemical Identifiers
7732-18-5 | |
B |
Specifications
Boron | |
B | |
Bottle | |
Liquid | |
Atomic Absorption Spectroscopy Flame Single-Element Standard Solution |
7732-18-5 | |
1g/L ±0.002g/L | |
H2O | |
100mL |