Få mere at vide
(+)-Bornyl Acetate, TRC
Artikelnummer. 30338543
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30338543 250 mg
1 options
Artikelnummer. 30338543 Leverandør Toronto Research Chemicals Leverandørnr. TRCB675535250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (+)-Bornyl Acetate
CAS 20347-65-3
Molekylær formel C12 H20 O2
Mængde 250 mg
Synonym Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, 2-acetate, (1R,2S,4R)-, 2-Bornanol, acetate, (1R,2S,4R)-(+)- (8CI), Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, acetate, (1R,2S,4R)- (9CI), Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, acetate, (1R-endo)-, Borneol, d-, acetate (6CI), (+)-Borneol acetate, (+)-Bornyl, acetate, d-Bornyl acetate
InChI formel InChI=1S/C12H20O2/c1-8(13)14-10-7-9-5-6-12(10,4)11(9,2)3/h9-10H,5-7H2,1-4H3/t9-,10+,12+/m1/s1
SMIL CC(=O)O[C@H]1C[C@H]2CC[C@]1(C)C2(C)C
IUPAC navn [(1R,3S,4R)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate
Molekylvægt (g/mol) 196.29
Formel vægt 196.1463