Få mere at vide
Bornyl Acetate (Mixture of Diastereomers), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bornyl Acetate |
| CAS | 76-49-3 |
| Renhedsgrad noter | GC |
| Molekylær formel | C12 H20 O2 |
| Mængde | 2.5 g |
| Synonym | Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, 2-acetate, (1R,2S,4R)-rel-, Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, acetate, (1R,2S,4R)-rel- (9CI), Bicyclo[2.2.1]heptan-2-ol, 1,7,7-trimethyl-, acetate, endo-, Borneol, acetate (6CI,7CI,8CI), (+/-)-Bornyl acetate, Borneyl acetate, Bornyl acetate, Bornyl acetic ester, Bornyl ethanoate, dl-Bornyl acetate, endo-Bornyl acetate |
| InChI formel | InChI=1S/C12H20O2/c1-8(13)14-10-7-9-5-6-12(10,4)11(9,2)3/h9-10H,5-7H2,1-4H3/t9-,10+,12+/m0/s1 |
| SMIL | CC(=O)O[C@@H]1C[C@@H]2CC[C@@]1(C)C2(C)C |
| IUPAC navn | [(1S,3R,4S)-4,7,7-trimethyl-3-bicyclo[2.2.1]heptanyl] acetate |
| Molekylvægt (g/mol) | 196.29 |
| Vis mere |