missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Borane-tert-butylamine complex, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Borane-tert-butylamine complex is used in stereoselective reduction of a steroidal ketone and for a review of amine boranes as selective reducing and hydroborating agents. It is also used as strong co-reducing agent in the syntheses of copper (I) oxide nanoparticles, diphosphine-protected gold nanocluster and 4-acetoxycinnamyl alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Borane-tert-butylamine complex is used in stereoselective reduction of a steroidal ketone and for a review of amine boranes as selective reducing and hydroborating agents. It is also used as strong co-reducing agent in the syntheses of copper (I) oxide nanoparticles, diphosphine-protected gold nanocluster and 4-acetoxycinnamyl alcohols.
Solubility
Soluble in water (27 g/L at 20°C) and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and acids.
Borane-tert-butylamine complex is used in stereoselective reduction of a steroidal ketone and for a review of amine boranes as selective reducing and hydroborating agents. It is also used as strong co-reducing agent in the syntheses of copper (I) oxide nanoparticles, diphosphine-protected gold nanocluster and 4-acetoxycinnamyl alcohols.
Solubility
Soluble in water (27 g/L at 20°C) and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and acids.
Specifications
Melting Point | 96°C to 98°C |
Density | 0.87 |
Quantity | 10 g |
UN Number | UN2811 |
Beilstein | 4134905 |
Solubility Information | Soluble in water (27g/L at 20°C) and methanol. |
Formula Weight | 86.98 |
Percent Purity | 97% |
Chemical Name or Material | Borane-tert-butylamine complex |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'