missing translation for 'onlineSavingsMsg'
Learn More

Boldenone Undecylenate 1.0 mg/ml in Acetonitrile, LoGiCal

Product Code. 30463928
Change view
Click to view available options
Mængde:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30463928 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463928 Supplier LoGiCal Supplier No. LGCAMP3560.0011

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Boldenone Undecylenate
CAS 13103-34-9
Analyt- eller komponentnavne Boldenone Undecylenate
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C30 H44 O3
Certificeringer/compliance ISO 17034
Synonym Ba 29038, Ba 9038, Boldefarm, Boldenone 10-undecenoate, Boldenone undecylenate, Equipoise, Parenabol, Vebonol, (17beta)-17-[(1-Oxo-10-undecenyl)oxy]-androsta-1,4-dien-3-one, Androsta-1,4-dien-3-one, 17beta-hydroxy-, 10-undecenoate (7CI,8CI), 10-Undecenoic acid, ester with 17beta-hydroxyandrosta-1,4-dien-3-one (8CI)
InChI formel InChI=1S/C30H44O3/c1-4-5-6-7-8-9-10-11-12-28(32)33-27-16-15-25-24-14-13-22-21-23(31)17-19-29(22,2)26(24)18-20-30(25,27)3/h4,17,19,21,24-27H,1,5-16,18,20H2,2-3H3/t24-,25-,26-,27-,29-,30-/m0/s1
Løsningstype Single Solution
SMIL C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCCCCCCCC=C
IUPAC navn [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] undec-10-enoate
Opløsningsmiddel eller Matrix Acetonitrile
Industry Type Food Testing
Molekylvægt (g/mol) 452.67
Mængde 1 ml
Formel vægt 452.329 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.