Learn More
Bohlender™ BOLA™ Spare V-rings, Two-part Set
Set of two spare v-rings.
Brand:  Bohlender™ D502-04
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 17177194
  • 157.94 DKK / Pack of 10
Estimated Shipment: 24-06-2024
to see stock.
Add to basket
Description
Description
  • Made of PTFE
  • Two-part set with one tapered ring and one v-ring
Specifications
Specifications
PTFE
8 mm
M 18 x 2
One Tapered Ring, One V-ring