missing translation for 'onlineSavingsMsg'
Få mere at vide

Bodipy 8-Chloromethane, TRC

Artikelnummer. 30329331
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30329331 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30329331 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6747005MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Bodipy 8-Chloromethane
CAS 208462-25-3
Molekylær formel C14H16BClF2N2
Mængde 5 mg
Synonym (T-4)-[2-[2-Chloro-1-(3,5-dimethyl-2H-pyrrol-2-ylidene-κN)ethyl]-3,5-dimethyl-1H-pyrrolato-κN]difluoro-boron
InChI formel InChI=1S/C14H16BClF2N2/c1-8-5-10(3)19-13(8)12(7-16)14-9(2)6-11(4)20(14)15(19,17)18/h5-6H,7H2,1-4H3
SMIL [B-]1(N2C(=CC(=C2C(=C3[N+]1=C(C=C3C)C)CCl)C)C)(F)F
IUPAC navn 8-(chloromethyl)-2,2-difluoro-4,6,10,12-tetramethyl-3-aza-1-azonia-2-boranuidatricyclo[7.3.0.03,7]dodeca-1(12),4,6,8,10-pentaene
Molekylvægt (g/mol) 296.55
Formel vægt 296.1063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.