Få mere at vide
Boceprevir-d9, TRC
Artikelnummer. 30331669
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30331669 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331669 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6745025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Boceprevir-d9
CAS 1256751-11-7
Molekylær formel C27 2H9 H36 N5 O5
Mængde 5 mg
Synonym Boceprevir D9
InChI formel InChI=1S/C27H45N5O5/c1-25(2,3)20(30-24(37)31-26(4,5)6)23(36)32-13-15-17(27(15,7)8)18(32)22(35)29-16(19(33)21(28)34)12-14-10-9-11-14/h14-18,20H,9-13H2,1-8H3,(H2,28,34)(H,29,35)(H2,30,31,37)/t15-,16?,17-,18-,20+/m0/s1/i4D3,5D3,6D3
SMIL [2H]C([2H])([2H])C(NC(=O)N[C@H](C(=O)N1C[C@H]2[C@@H]([C@H]1C(=O)NC(CC3CCC3)C(=O)C(=O)N)C2(C)C)C(C)(C)C)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn (1R,2S,5S)-N-(4-amino-1-cyclobutyl-3,4-dioxobutan-2-yl)-3-[(2S)-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]carbamoylamino]-3,3-dimethylbutanoyl]-6,6-dimethyl-3-azabicyclo[3.1.0]hexane-2-carboxamide
Molekylvægt (g/mol) 528.73
Formel vægt 528.3986