Få mere at vide
Boc-trans-4-hydroxy-D-proline, TRC
Artikelnummer. 30135861
Skift visning
Mængde:
500 mg
1 g
2 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135861 1 g
30132866 2 g
30012418 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135861 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2849401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 147266-92-0
Molekylær formel C10H17F2NO5
Molekylvægt (g/mol) 231.25
Synonym (2R,4S)-1-(Tert-Butoxycarbonyl)-4-Hydroxypyrrolidine-2-Carboxylic Acid
IUPAC navn (2R,4S)-4-hydroxy-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMIL CC(C)(C)OC(=O)N1C[C@@H](O)C[C@@H]1C(=O)O
Tekniske data
Kemisk navn eller materiale Boc-trans-4-hydroxy-D-proline
InChI formel InChI=1S/C10H17NO5/c1-10(2,3)16-9(15)11-5-6(12)4-7(11)8(13)14/h6-7,12H,4-5H2,1-3H3,(H,13,14)/t6-,7+/m0/s1
Mængde 1 g