Learn More
BOC-N-methyl-D-alanine, 97%, Thermo Scientific™
Catalog Number p-4228426
Click to view available options
Quantity:
1 g
5 g
19914-38-6
C9H17NO4
203.24
MFCD00063137
VLHQXRIIQSTJCQ-ZCFIWIBFSA-N
boc-n-methyl-d-alanine, boc-n-me-d-ala-oh, n-boc-n-methyl-d-alanine, boc-d-n-me-ala-oh, boc-n-alpha-methyl-d-alanine, r-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-d-alanine, d-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2r-2-tert-butoxycarbonyl methyl amino propanoic acid
16211306
(2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
CC(C(=O)O)N(C)C(=O)OC(C)(C)C
19914-38-6
C9H17NO4
203.24
MFCD00063137
VLHQXRIIQSTJCQ-ZCFIWIBFSA-N
boc-n-methyl-d-alanine, boc-n-me-d-ala-oh, n-boc-n-methyl-d-alanine, boc-d-n-me-ala-oh, boc-n-alpha-methyl-d-alanine, r-2-tert-butoxycarbonyl methyl amino propanoic acid, n-tert-butoxycarbonyl-n-methyl-d-alanine, d-alanine, n-1,1-dimethylethoxy carbonyl-n-methyl, 2r-2-tert-butoxycarbonyl methyl amino propanoic acid
16211306
(2R)-2-[methyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]propanoic acid
CC(C(=O)O)N(C)C(=O)OC(C)(C)C
Melting Point 92.3°C to 94°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation 29° to 31° (20°C, 589nm) (c=1, ethanol)
Formula Weight 203.24
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material BOC-N-Methyl-D-alanine