missing translation for 'onlineSavingsMsg'
Learn More

BOC-D-α-phenylglycine, 99%, Thermo Scientific Chemicals

Catalog Number p-4228423
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
33125-05-2
C13H17NO4
251.28
MFCD00062043,MFCD00065588
HOBFSNNENNQQIU-UHFFFAOYNA-N
boc-d-phg-oh, boc-d-phenylglycine, n-boc-d-phenylglycine, n-boc-d-2-phenylglycine, n-boc-l-phenylglycine, n-tert-butoxycarbonyl-d-2-phenylglycine, boc-d-alpha-phenylglycine, r-tert-butoxycarbonyl amino phenyl acetic acid, r-tert-butoxycarbonylamino-phenyl-acetic acid, r-2-tert-butoxycarbonyl amino-2-phenylacetic acid
2755953
CC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1
This item is not returnable. View return policy
33125-05-2
C13H17NO4
251.28
MFCD00062043,MFCD00065588
HOBFSNNENNQQIU-UHFFFAOYNA-N
boc-d-phg-oh, boc-d-phenylglycine, n-boc-d-phenylglycine, n-boc-d-2-phenylglycine, n-boc-l-phenylglycine, n-tert-butoxycarbonyl-d-2-phenylglycine, boc-d-alpha-phenylglycine, r-tert-butoxycarbonyl amino phenyl acetic acid, r-tert-butoxycarbonylamino-phenyl-acetic acid, r-2-tert-butoxycarbonyl amino-2-phenylacetic acid
2755953
CC(C)(C)OC(=O)NC(C(O)=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 88°C to 92°C
Color White
Assay Percent Range 98.5% min. (HPLC)
Infrared Spectrum Authentic
Packaging Glass Bottle
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid
Specific Rotation -145
Formula Weight 251.28
Percent Purity 99%
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material BOC-D-α-phenylglycine
Show More Show Less
compliance-icons
Product Identifier
  • N-BOC-D-Phenylglycine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'