missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Boc-aminooxy)acetic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
(Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
(Boc-aminooxy)acetic acid was employed to introduce a hydroxylamine moiety into peptides; reaction with an aldehyde to an oxime. It was used in the preparation of Boc-aminooxy tetra(ethylene glycol).
Solubility
Soluble in methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | ∽115°C (decomposition) |
Linear Formula | (CH3)3COCONHOCH2CO2H |
Quantity | 1 g |
Beilstein | 6137646 |
Solubility Information | Soluble in methanol. |
Formula Weight | 191.18 |
Percent Purity | ≥98% |
Chemical Name or Material | (Boc-aminooxy)acetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'