Få mere at vide
Invitrogen™ Human BMX, His Tag Recombinant Protein
Beskrivelse
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length, Recombinant, Cytoplasmic.
Tekniske data
Tekniske data
| Adgangsnummer | P51813 |
| Koncentration | See Label |
| Til brug med (applikation) | Kinase Assay |
| Formulering | 50mM tris HCl with 4mM DTT, 0.05% Triton X-100, 50% glycerol, 150mM NaCl, 0.5mM EDTA and no preservative; pH 7.5 |
| Gen-id (Entrez) | 660 |
| Molekylvægt (g/mol) | 83 kDa |
| Navn | Human BMX, His Tag |
| pH-område | 7.5 |
| Oprensningsmetode | Purified |
| Mængde | 100 μg |
| Vis mere |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.