Få mere at vide
BMS 564929, Tocris Bioscience™
Highly potent and muscle-selective androgen receptor (AR) agonist
Brand: Tocris Bioscience 5274/5
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
BMS 564929 is a highly potent and selective androgen receptor (AR) agonist (EC50 = 0.44 nM). Exhibits selectivity for ARs over estrogen, glucocorticoid and mineralocorticoid receptors and no significant binding to steroid hormone binding globulin or aromatase. Also exhibits selectivity for muscle over prostate ARs. Active in vivo and orally bioavailable.Tekniske data
| 2-Chloro-3-methyl-4-[(7R,7aS)-tetrahydro-7-hydroxy-1,3-dioxo-1H-pyrrolo[1,2-c]imidazol-2(3H)-yl]benzonitrile | |
| 627530-84-1 | |
| >98% | |
| 5 mg |