missing translation for 'onlineSavingsMsg'
Få mere at vide

STEMCELL Technologies BMS 493

Artikelnummer. 17140302
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17140302 1 mg
17150302 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17140302 Leverandør STEMCELL Technologies Leverandørnr. 73972

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Retinoid pathway activator; Activates pan-retinoic acid receptor

  • BMS 493 is an inverse agonist of the pan-retinoic acid receptors (RARs)
  • RARs are ligand-dependent transcription factors that regulate gene networks that control cell growth, differentiation, survival, and death (Germain et al.) BMS 493 has been shown to increase nuclear receptor co-repressor (NCoR) interaction with RARs (Germain et al.; Rinkevich et al.)
  • Affects development of bronchial tubule formation in mice (Chazaud et al.)
  • Inhibits activation of immature dendritic cells (Geissmann et al.)
  • Inhibits differentiation of leukemic blasts (Kamashev et al.)

Tekniske data

CAS 215030-90-3
Synonym BMS 204493
Mængde 1 mg
Molekylær formel C29H24O2
Renhed ≥98%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.