missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BMPR1A (ALK3), GST Tag Recombinant Protein
Human BMPR1A (ALK3), GST Tag Recombinant Protein
Brand: Invitrogen™ PR9136A
Additional Details : Weight : 0.01000kg
Description
BMPR1A phosphorylates intracellular SMADs in response to bone morphogenic proteins.
Specifications
NP_004320.2 | |
67kDa | |
Kinase proteins are shipped on dry ice and should be stored at -80°C. | |
Catalytic | |
ALK Kinase Family | |
Recombinant | |
Insect |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
100μg | |
RUO - research use only | |
BMPR1A (ALK3) | |
Human | |
GST Tag | |
Frozen |