Få mere at vide
Brady™ BMP61 BMP71 M611 Polyester Labels
Farve:
Grå
Hvid
Klarhed
Sølv
Dimensioner (LxB):
15 mx 4,5 mm
15 mx 8 mm
15,24 m x 48,26 mm
15,54 m x 48,26 mm
Beskrivelse
- Material is UL approved with its respective ribbon
- See the Recommended Ribbon Series for the appropriate ribbon to use
- Can also be printed on the BMP™71 Label Printer provided the roll adapter (M71-ADAPT) is used
Tekniske data
Tekniske data
| Klæbende type | Permanent Acrylic |
| Farve | Hvid |
| Slutte | Gloss |
| Materiale | Polyester (B-422) |
| Print teknologi | Thermal Transfer |
| Til brug med (udstyr) | BMP61, BMP71, M611 |
| Dimensioner (LxB) | 15 mx 8 mm |