missing translation for 'onlineSavingsMsg'
Learn More
BML-284, MedChemExpress Product Code.: 16473927

BML-284, MedChemExpress

Product Code. p-200100042
50 mg, 50mg
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473927

Brand: Medchem Express HY19987

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BML-284 is a potent and cell-permeable Wnt signaling activator. BML-284 induces TCF-dependent transcriptional activity with an EC50 of 700 nM.

BML-284 is a potent and cell-permeable Wnt signaling activator. BML-284 induces TCF-dependent transcriptional activity with an EC50 of 700 nM.

Specifications

Chemical Name or Material BML-284
CAS 853220-52-7
Color White
Purity Grade Notes Research
Molecular Formula C19H18N4O3
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (285.41 mM)
SMILES NC1=NC(C2=CC=CC(OC)=C2)=CC(NCC3=CC=C(OCO4)C4=C3)=N1
Molecular Weight (g/mol) 350.37
Formula Weight 350.37
Percent Purity 97.48%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
BML-284, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.