Få mere at vide
BML-277, MedChemExpress
Artikelnummer. 16435928
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16435928 10 mg
16422077 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16435928 Leverandør Medchem Express Leverandørnr. HY13946
MedChemExpress BML-277 is a selective checkpoint kinase 2 (Chk2) inhibitor with an IC50 of 15 nM.
BML-277 is a selective checkpoint kinase 2 (Chk2) inhibitor with an IC50 of 15 nM.
Tekniske data
Kemisk navn eller materiale BML-277
CAS 516480-79-8
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C20H14ClN3O2
Mængde 10 mg
Synonym Chk2 Inhibitor II
Opløselighedsinformation DMSO : 22.22 mg/mL (61.08 mM; Need ultrasonic)
SMIL ClC(C=C1)=CC=C1OC(C=C2)=CC=C2C3=NC4=CC(C(N)=O)=CC=C4N3
Molekylvægt (g/mol) 363.8
Formel vægt 363.8
Procent renhed 97.94%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease