missing translation for 'onlineSavingsMsg'
Learn More

Blonanserin N-Oxide, TRC

Product Code. 30341962
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30341962 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341962 Supplier Toronto Research Chemicals Supplier No. TRCB59585310MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Blonanserin N-Oxide
CAS 142838-82-2
Molekylær formel C23H30FN3O
Mængde 10 mg
Synonym 2-(4-Ethyl-4-oxido-1-piperazinyl)-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine, N-Oxide 2-(4-Ethyl-1-piperazinyl)-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine
InChI formel InChI=1S/C23H30FN3O/c1-2-27(28)15-13-26(14-16-27)23-17-21(18-9-11-19(24)12-10-18)20-7-5-3-4-6-8-22(20)25-23/h9-12,17H,2-8,13-16H2,1H3
SMIL FC(C=C1)=CC=C1C2=CC(N3CCN(CC3)(CC)=O)=NC4=C2CCCCCC4
IUPAC navn 2-(4-ethyl-4-oxidopiperazin-4-ium-1-yl)-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine
Molekylvægt (g/mol) 383.5
Formel vægt 383.2373
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.