missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ BLCAP Monoclonal Antibody (4E1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA532921
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
BLCAP Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a tumor suppressor protein that reduces cell growth by stimulating apoptosis. The first intron of this gene contains the neuronatin gene, which is imprinted and only expressed from the paternal allele. This gene is imprinted in brain where it is differentially expressed from different promoters. Transcription from the upstream promoter occurs preferentially on the maternal allele, and transcripts are preferentially expressed from the downstream promoter on the paternal allele. Alternative promoters and alternative splicing result in multiple transcript variants that encode the same protein.[provided by RefSeq, Nov 2009].
Tekniske data
| BLCAP | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 50% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P62950, P62952 | |
| BLCAP | |
| Synthetic peptide within Human BLCAP aa 38-87. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 4E1 | |
| Unconjugated | |
| BLCAP | |
| AI462828; Bc10; bladder cancer 10 kDa protein; bladder cancer associated protein; bladder cancer associated protein homolog; bladder cancer associated protein homolog (human); bladder cancer related protein; bladder cancer related protein (10kD); bladder cancer-associated protein; BLCAP; Blcapbladder cancer associated protein homolog | |
| Mouse | |
| Protein G | |
| RUO | |
| 10904, 171113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion