Learn More
BRAND™ BLAUBRAND™ Bulb Pipettes 1-Mark
Class AS, 1-Mark Bulb Pipettes offer higher measurement accuracy than graduated pipettes.
Brand: BRAND™ 230608
561.26 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- AR-Glas™
- Class 1-mark graduated
- Includes batch certificate of conformity
Specifications
Class AS | |
1-Mark Graduations | |
6 mL |
AR-GLAS | |
Batch Certificate of Conformity | |
Bulb Pipet |
Product Content Correction
Product Title
BRAND™ BLAUBRAND™ Bulb Pipettes 1-Mark > Bulb Pipette; Volume (Metric): 6 mL
Spot an opportunity for improvement?Share a Content Correction