missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ Black Cyclopore™ Membranes
For use with epifluorescence microscopy and applications requiring contrasting background.
Brand: Cytiva 7063-4702
Additional Details : Weight : 0.06000kg
Description
For use with epifluorescence microscopy and applications requiring contrasting background.
- Black polycarbonate membrane
- Dark membrane reduces background fluorescence
- Improves sensitivity of test
- Membrane used to filter test and then used for analysis
Specifications
Autoclavable | |
Membrane Filter | |
0.2 μm | |
Hydrophilic | |
47 mm |
Black | |
Polycarbonate | |
Highly suitable for epifluorescence and other microscopy applications | |
Filtration Membrane Disc | |
47 mm |